siRNA Details
Gene Symbol: Txndc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005559.2 | NP_001005559.2 | 2 | 1269 | ||
NM_001146004.1 | NP_001139476.1 | 2 | 1447 | |||
XM_006245607.2 | XP_006245669.1 | 3 | 1321 | |||
XM_008767379.1 | XP_008765601.1 | 3 | 1844 | |||
GenBank mRNA | BC079238.1 | - | 2 | 1447 | 7132630 | |
BC083924.1 | - | 3 | 1327 | 7135313 |
