siRNA Details
Gene Symbol: Dhrs11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014119.2 | NP_001014141.2 | 4 | 611 | ||
XM_006247045.2 | XP_006247107.1 | 4 | 732 | |||
GenBank mRNA | BC088444.1 | - | 4 | 605 | 7126427 | |
DY311021.1 | - | 4 | 611 |
