siRNA Details
Gene Symbol: LOC685157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115044.1 | NP_001108516.1 | 2 | 241 | ||
XM_008769100.1 | XP_008767322.1 | 2 | 293 | |||
GenBank mRNA | BC158830.1 | - | 2 | 241 | 7379096 | |
FQ227411.1 | - | 1 | 176 |
Gene Symbol: LOC685438 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769097.1 | XP_008767319.1 | 2 | 190 |
