siRNA Details
Gene Symbol: Gatc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108339.1 | NP_001101809.1 | 363 | |||
XM_008769339.1 | XP_008767561.1 | 401 |
Gene Symbol: LOC100909836 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751199.3 | XP_003751247.1 | 2 | 401 |
