siRNA Details
Gene Symbol: Ube2t | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108344.2 | NP_001101814.2 | 2, 3 | 274 | ||
GenBank mRNA | AW141189.1 | - | 2, 3 | 274 | ||
BC167012.1 | - | 2, 3 | 235 | 9099623 | ||
FQ233263.1 | - | 2, 3 | 262 |
