siRNA Details
Gene Symbol: RGD1566099 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108347.1 | NP_001101817.1 | 2 | 299 | ||
GenBank mRNA | BC166874.1 | - | 2 | 296 | 6890641 | |
BC166876.1 | - | 2 | 296 | 6919569 | ||
BC166924.1 | - | 2 | 296 | 6919449 |
