siRNA Details
Gene Symbol: Dok4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108438.2 | NP_001101908.1 | 9 | 1205 | ||
XM_008772314.1 | XP_008770536.1 | 7 | 1297 | |||
XM_008772315.1 | XP_008770537.1 | 8 | 1389 | |||
XM_008772316.1 | XP_008770538.1 | 8 | 1386 | |||
XM_008772317.1 | XP_008770539.1 | 8 | 1145 | |||
GenBank mRNA | BC158749.1 | - | 9 | 1205 | 9101502 |
