siRNA Details
Gene Symbol: Dok4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108438.2 | NP_001101908.1 | 5 | 606 | ||
XM_008772314.1 | XP_008770536.1 | 4 | 547 | |||
XM_008772315.1 | XP_008770537.1 | 5 | 639 | |||
XM_008772316.1 | XP_008770538.1 | 5 | 636 | |||
XM_008772317.1 | XP_008770539.1 | 4 | 546 | |||
GenBank mRNA | BC158749.1 | - | 5 | 606 | 9101502 |
