siRNA Details
Gene Symbol: Ube3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191837.2 | NP_001178766.1 | 5 | 863 | ||
XM_006229310.2 | XP_006229372.1 | 4 | 1708 | |||
XM_006229313.2 | XP_006229375.1 | 3 | 1588 | |||
XM_008759399.1 | XP_008757621.1 | 2 | 2418 | |||
XM_008759400.1 | XP_008757622.1 | 4 | 349 | |||
XM_008759401.1 | XP_008757623.1 | 4 | 1652 |
