siRNA Details
Gene Symbol: LOC100360933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193275.1 | NP_001180204.1 | 5 | 553 | ||
XM_008759490.1 | XP_008757712.1 | 2 | 921 | |||
XM_008759491.1 | XP_008757713.1 | 2 | 409 | |||
GenBank mRNA | BF386180.1 | - | 2 | 161 |
