siRNA Details
Gene Symbol: LOC100360933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193275.1 | NP_001180204.1 | 6 | 614 | ||
XM_008759491.1 | XP_008757713.1 | 3 | 470 | |||
GenBank mRNA | BF386180.1 | - | 1 | 100 |
