siRNA Details
Gene Symbol: Relt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108495.1 | NP_001101965.1 | 4 | 370 | ||
XM_006229775.2 | XP_006229837.1 | 5 | 538 | |||
XM_006229776.2 | XP_006229838.1 | 5 | 2802 | |||
XM_006229777.2 | XP_006229839.1 | 5 | 489 | |||
XM_006229778.2 | XP_006229840.1 | 5 | 4890 | |||
XM_006229780.2 | XP_006229842.1 | 5 | 538 | |||
XM_008759763.1 | XP_008757985.1 | 5 | 5218 | |||
XM_008759764.1 | XP_008757986.1 | 5 | 5143 | |||
XM_008759765.1 | XP_008757987.1 | 5 | 5098 |
