siRNA Details
Gene Symbol: RGD1563222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108585.1 | NP_001102055.1 | 2 | 607 | ||
XM_008762062.1 | XP_008760284.1 | 4 | 5695 | |||
XM_008762063.1 | XP_008760285.1 | 4 | 4791 | |||
XM_008762064.1 | XP_008760286.1 | 5 | 5366 | |||
XM_008762065.1 | XP_008760287.1 | 5 | 5563 | |||
XM_008762066.1 | XP_008760288.1 | 4 | 3554 |
