siRNA Details
Gene Symbol: Tbc1d20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004281.1 | NP_001004281.1 | 7 | 942 | ||
XM_008762348.1 | XP_008760570.1 | 7 | 1028 | |||
GenBank mRNA | BC079383.1 | - | 7 | 942 | 7111239 |
