siRNA Details
Gene Symbol: Snta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100901.1 | NP_001094371.1 | 2 | 506 | ||
XM_006235326.2 | XP_006235388.1 | 2 | 545 | |||
GenBank mRNA | BC169101.1 | - | 2 | 506 | 5599966 |
