siRNA Details
Gene Symbol: Gpr162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108646.1 | NP_001102116.1 | 2 | 716 | ||
XM_006237371.2 | XP_006237433.1 | 2 | 813 |

More Information
Other Species Targeted

Gene Ontology Categories:
