siRNA Details
Gene Symbol: Rbks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108703.1 | NP_001102173.1 | 5 | 420 | ||
XM_006239806.2 | XP_006239868.1 | 5 | 466 | |||
XM_008764520.1 | XP_008762742.1 | 5 | 914 | |||
XM_008764521.1 | XP_008762743.1 | 5 | 466 | |||
GenBank mRNA | FQ209797.1 | - | 5 | 402 |
