siRNA Details
Gene Symbol: Mum1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108736.2 | NP_001102206.2 | 13 | 2072 | ||
XM_006240973.2 | XP_006241035.1 | 13 | 2108 | |||
XM_006240974.2 | XP_006241036.1 | 13 | 2141 | |||
XM_006240975.2 | XP_006241037.1 | 13 | 3062 | |||
GenBank mRNA | BC160828.1 | - | 13 | 2075 | 7453186 | |
CK839769.1 | - | 2 | 388 |
