siRNA Details
Gene Symbol: Cd320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014201.1 | NP_001014223.1 | 5 | 746 | ||
GenBank mRNA | BC088861.1 | - | 5 | 746 | 7310701 | |
FQ219198.1 | - | 5 | 748 | |||
FQ234194.1 | - | 5 | 759 |
