siRNA Details
Gene Symbol: Elk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108743.1 | NP_001102213.1 | 2 | 552 | ||
XM_006241265.2 | XP_006241327.1 | 3 | 957 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013508, S82864, BC053402, Z32815, BC055735, and others.
Gene Ontology Categories:
