siRNA Details
Gene Symbol: Tmem184b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173370.1 | NP_001166841.1 | 6 | 746 | ||
XM_006242013.2 | XP_006242075.1 | 6 | 753 | |||
XM_006242014.2 | XP_006242076.1 | 6 | 1339 | |||
GenBank mRNA | BC099785.1 | - | 6 | 746 |
