siRNA Details
Gene Symbol: Rangap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244855.1 | NP_001231784.1 | 4 | 477 | ||
XM_006242100.2 | XP_006242162.1 | 4 | 1917 | |||
XM_006242101.2 | XP_006242163.1 | 4 | 766 | |||
GenBank mRNA | BC082056.1 | - | 4 | 455 | 7132303 | |
CV126932.1 | - | 4 | 477 | |||
FQ217036.1 | - | 4 | 461 |
