siRNA Details
Gene Symbol: Rangap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244855.1 | NP_001231784.1 | 6 | 786 | ||
XM_006242100.2 | XP_006242162.1 | 6 | 2226 | |||
XM_006242101.2 | XP_006242163.1 | 6 | 1075 | |||
GenBank mRNA | BC082056.1 | - | 6 | 764 | 7132303 | |
DY317647.1 | - | 2 | 258 | |||
FQ217036.1 | - | 6 | 770 |
