siRNA Details
Gene Symbol: RGD1304694 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271047.1 | NP_001257976.1 | 6 | 782 | ||
GenBank mRNA | BC098850.1 | - | 1 | 56 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1304694 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271047.1 | NP_001257976.1 | 6 | 782 | ||
GenBank mRNA | BC098850.1 | - | 1 | 56 |