siRNA Details
Gene Symbol: Tarbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034941.1 | NP_001030113.1 | 8 | 1019 | ||
XM_006242428.2 | XP_006242490.1 | 8 | 1099 | |||
XM_006242429.2 | XP_006242491.1 | 7 | 912 | |||
XM_006242430.2 | XP_006242492.1 | 6 | 757 | |||
GenBank mRNA | BC104690.1 | - | 8 | 1019 | 7107060 | |
FQ228813.1 | - | 8 | 1016 |
