siRNA Details
Gene Symbol: Dyx1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007010.1 | NP_001007011.1 | 9 | 1252 | ||
XM_006243328.2 | XP_006243390.1 | 9 | 1321 | |||
XM_006243329.2 | XP_006243391.1 | 8 | 1226 | |||
XM_006243331.2 | XP_006243393.1 | 7 | 972 | |||
XM_008766313.1 | XP_008764535.1 | 9 | 1230 | |||
XM_008766314.1 | XP_008764536.1 | 9 | 1061 | |||
GenBank mRNA | AY428506.1 | - | 9 | 1252 | ||
BC085838.1 | - | 9 | 1196 | 7135744 |
