siRNA Details
Gene Symbol: Rassf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007754.1 | NP_001007755.1 | 3 | 321 | ||
NM_001037555.1 | NP_001032644.1 | 4 | 513 | |||
GenBank mRNA | AB202124.1 | - | 4 | 513 | ||
BC079237.1 | - | 3 | 321 | 7132629 | ||
FQ233012.1 | - | 3 | 361 |
