siRNA Details
Gene Symbol: Pde6d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108806.1 | NP_001102276.1 | 5 | 552 | ||
XM_006245413.2 | XP_006245475.1 | 5 | 600 | |||
GenBank mRNA | FQ216262.1 | - | 3 | 174 | ||
FQ227000.1 | - | 3 | 174 | |||
FQ234983.1 | - | 3 | 174 |
