siRNA Details
Gene Symbol: Atg16l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108809.3 | NP_001102279.2 | 17 | 1708 | ||
XM_006245419.2 | XP_006245481.1 | 18 | 1934 | |||
XM_006245420.2 | XP_006245482.1 | 16 | 1828 | |||
GenBank mRNA | CK473997.1 | - | 1 | 180 | ||
CK481169.1 | - | 5 | 427 |
