siRNA Details
Gene Symbol: Stac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108834.1 | NP_001102304.1 | 1 | 497 | ||
XM_006247383.2 | XP_006247445.1 | 1 | 388 | |||
GenBank mRNA | AM903074.1 | - | 1 | 22 |
