siRNA Details
Gene Symbol: Asphd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009716.1 | NP_001009716.1 | 2, 3 | 1359 | ||
XM_008769345.1 | XP_008767567.1 | 3, 4 | 2592 | |||
XM_008769346.1 | XP_008767568.1 | 4, 5 | 2822 | |||
XM_008769347.1 | XP_008767569.1 | 4, 5 | 1782 | |||
XM_008769348.1 | XP_008767570.1 | 4, 5 | 2495 | |||
XM_008769349.1 | XP_008767571.1 | 4, 5 | 2378 | |||
XM_008769350.1 | XP_008767572.1 | 3, 4 | 2280 | |||
XM_008769351.1 | XP_008767573.1 | 2, 3 | 1238 | |||
GenBank mRNA | BC088863.1 | - | 2, 3 | 1359 | 7313227 |
