siRNA Details
Gene Symbol: Asphd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009716.1 | NP_001009716.1 | 2 | 1081 | ||
XM_008769345.1 | XP_008767567.1 | 3 | 2314 | |||
XM_008769346.1 | XP_008767568.1 | 4 | 2544 | |||
XM_008769347.1 | XP_008767569.1 | 4 | 1504 | |||
XM_008769348.1 | XP_008767570.1 | 4 | 2217 | |||
XM_008769349.1 | XP_008767571.1 | 4 | 2100 | |||
XM_008769350.1 | XP_008767572.1 | 3 | 2002 | |||
XM_008769351.1 | XP_008767573.1 | 2 | 960 | |||
GenBank mRNA | BC088863.1 | - | 2 | 1081 | 7313227 |
