siRNA Details
Gene Symbol: Syce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191557.1 | NP_001178486.1 | 2 | 255 | ||
XM_006255291.2 | XP_006255353.1 | 3 | 1805 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Syce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191557.1 | NP_001178486.1 | 2 | 255 | ||
XM_006255291.2 | XP_006255353.1 | 3 | 1805 |