siRNA Details
Gene Symbol: Lyrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276487.1 | NP_001263416.1 | 4 | 443 | ||
XM_006230132.2 | XP_006230194.1 | 3 | 383 | |||
XM_006230133.1 | XP_006230195.1 | 3 | 379 | |||
XM_006230134.2 | XP_006230196.2 | 4 | 2686 | |||
XM_008759783.1 | XP_008758005.1 | 3 | 2593 | |||
GenBank mRNA | BC166909.1 | - | 4 | 440 | 6889767 |
