siRNA Details
Gene Symbol: Rpl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031099.1 | NP_112361.1 | 5 | 729 | ||
XM_006250585.2 | XP_006250647.1 | 5 | 710 | |||
GenBank mRNA | BC060561.1 | - | 5 | 676 | 6918805 | |
FQ212710.1 | - | 5 | 686 | |||
FQ214281.1 | - | 5 | 676 | |||
FQ214493.1 | - | 5 | 677 | |||
FQ215486.1 | - | 4 | 550 | |||
FQ219244.1 | - | 5 | 675 | |||
FQ219787.1 | - | 5 | 677 | |||
FQ219848.1 | - | 5 | 677 | |||
FQ220068.1 | - | 5 | 677 | |||
FQ220161.1 | - | 5 | 677 | |||
FQ220170.1 | - | 5 | 677 | |||
FQ220205.1 | - | 5 | 677 | |||
FQ220219.1 | - | 5 | 677 | |||
FQ227439.1 | - | 5 | 677 | |||
FQ228473.1 | - | 5 | 677 | |||
FQ229451.1 | - | 5 | 678 | |||
FQ229816.1 | - | 5 | 675 | |||
FQ229964.1 | - | 5 | 679 | |||
FQ229979.1 | - | 5 | 677 | |||
FQ230017.1 | - | 5 | 675 | |||
FQ230061.1 | - | 5 | 680 | |||
FQ230102.1 | - | 5 | 677 | |||
FQ230128.1 | - | 5 | 705 | |||
FQ230161.1 | - | 5 | 703 | |||
FQ230794.1 | - | 5 | 680 | |||
FQ231729.1 | - | 5 | 675 | |||
FQ232540.1 | - | 5 | 677 | |||
FQ233314.1 | - | 5 | 702 | |||
FQ234171.1 | - | 5 | 707 | |||
M17419.1 | - | 5 | 662 | |||
X06148.1 | - | 5 | 729 |
