siRNA Details
Gene Symbol: Rab22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108966.1 | NP_001102436.1 | 5 | 622 | ||
XM_006235705.2 | XP_006235767.1 | 6 | 798 | |||
XM_008762529.1 | XP_008760751.1 | 6 | 798 | |||
GenBank mRNA | BC158650.1 | - | 6 | 622 | 8367507 |
