siRNA Details
Gene Symbol: LOC102551192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773038.1 | XP_008771260.1 | 1 | 83 |
Gene Symbol: LOC102551397 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773039.1 | XP_008771261.1 | 83 |
