siRNA Details
Gene Symbol: Hpse2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135762.1 | NP_001129234.1 | 1 | 250 | ||
XM_008760445.1 | XP_008758667.1 | 1 | 2434 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hpse2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135762.1 | NP_001129234.1 | 1 | 250 | ||
XM_008760445.1 | XP_008758667.1 | 1 | 2434 |