siRNA Details
Gene Symbol: Olr964 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001073.1 | NP_001001073.1 | 1 | 496 |
Gene Symbol: LOC100910607 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240817.1 | XP_006240879.1 | 2 | 274 |
