siRNA Details
Gene Symbol: Ctsql2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002813.2 | NP_001002813.2 | 3 | 299 | ||
XM_008771444.1 | XP_008769666.1 | 3 | 283 | |||
GenBank mRNA | BC097257.1 | - | 3 | 299 | 7460188 |
