siRNA Details
Gene Symbol: Fut8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002289.1 | NP_001002289.1 | 1 | 84 | ||
XM_008764825.1 | XP_008763047.1 | 4 | 592 | |||
XM_008764826.1 | XP_008763048.1 | 4 | 1512 | |||
GenBank mRNA | AJ781406.1 | - | 1 | 84 | ||
FN668936.1 | - | 4 | 592 | |||
FN668937.1 | - | 4 | 621 | |||
FN668938.1 | - | 1 | 306 |
