siRNA Details
Gene Symbol: Dpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105729.3 | NP_001099199.2 | 9, 10 | 976 | ||
XM_006228677.2 | XP_006228739.1 | 9, 10 | 1013 | |||
XM_006228678.2 | XP_006228740.1 | 9, 10 | 1010 | |||
XM_006228679.2 | XP_006228741.1 | 9, 10 | 1010 | |||
XM_006228680.2 | XP_006228742.1 | 8, 9 | 964 | |||
XM_006228681.2 | XP_006228743.1 | 8, 9 | 960 | |||
XM_006228682.2 | XP_006228744.1 | 8, 9 | 927 | |||
XM_006228684.2 | XP_006228746.1 | 7, 8 | 1571 | |||
XM_006228685.1 | XP_006228747.1 | 9, 10 | 1837 | |||
XM_008759180.1 | XP_008757402.1 | 9, 10 | 999 | |||
GenBank mRNA | BC159415.1 | - | 9, 10 | 904 | 8368092 | |
X66022.1 | - | 9, 10 | 1837 |
