siRNA Details
Gene Symbol: Dpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105729.3 | NP_001099199.2 | 6 | 657 | ||
XM_006228677.2 | XP_006228739.1 | 6 | 694 | |||
XM_006228678.2 | XP_006228740.1 | 6 | 691 | |||
XM_006228679.2 | XP_006228741.1 | 6 | 691 | |||
XM_006228680.2 | XP_006228742.1 | 6 | 696 | |||
XM_006228681.2 | XP_006228743.1 | 6 | 692 | |||
XM_006228682.2 | XP_006228744.1 | 5 | 608 | |||
XM_006228684.2 | XP_006228746.1 | 5 | 1303 | |||
XM_006228685.1 | XP_006228747.1 | 6 | 1518 | |||
XM_008759180.1 | XP_008757402.1 | 6 | 680 | |||
GenBank mRNA | BC159415.1 | - | 6 | 585 | 8368092 | |
CO396551.1 | - | 6 | 657 | |||
X66022.1 | - | 6 | 1518 |
