siRNA Details
Gene Symbol: Dpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105729.3 | NP_001099199.2 | 5, 6 | 589 | ||
XM_006228677.2 | XP_006228739.1 | 5, 6 | 626 | |||
XM_006228678.2 | XP_006228740.1 | 5, 6 | 623 | |||
XM_006228679.2 | XP_006228741.1 | 5, 6 | 623 | |||
XM_006228680.2 | XP_006228742.1 | 5, 6 | 628 | |||
XM_006228681.2 | XP_006228743.1 | 5, 6 | 624 | |||
XM_006228685.1 | XP_006228747.1 | 5, 6 | 1450 | |||
XM_008759180.1 | XP_008757402.1 | 5, 6 | 612 | |||
GenBank mRNA | BC159415.1 | - | 5, 6 | 517 | 8368092 | |
CO396551.1 | - | 5, 6 | 589 | |||
X66022.1 | - | 5, 6 | 1450 |
