siRNA Details
Gene Symbol: Gja5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019280.1 | NP_062153.1 | 2 | 884 | ||
XM_008761328.1 | XP_008759550.1 | 2 | 4023 | |||
GenBank mRNA | AF021806.1 | - | 2 | 884 | ||
BC070935.1 | - | 2 | 906 | 7097997 | ||
M83092.1 | - | 1 | 794 |
