siRNA Details
Gene Symbol: Cacna1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019294.2 | NP_062167.2 | 32 | 4609 | ||
XM_006250069.2 | XP_006250131.1 | 35 | 5983 | |||
GenBank mRNA | AY029412.1 | - | 32 | 4609 | ||
L15453.1 | - | 33 | 4706 |
