siRNA Details
Gene Symbol: Rgs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019341.1 | NP_062214.1 | 4 | 284 | ||
GenBank mRNA | AF241259.1 | - | 4 | 284 | ||
EF568934.1 | - | 4 | 348 | |||
FQ234716.1 | - | 4 | 401 |
