siRNA Details
Gene Symbol: Vapb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021847.3 | NP_068619.1 | 6 | 688 | ||
XM_006235706.2 | XP_006235768.2 | 6 | 862 | |||
GenBank mRNA | AF086631.1 | - | 6 | 688 | ||
BC065576.1 | - | 6 | 873 | 5598396 |
