siRNA Details
Gene Symbol: Cyb5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022245.1 | NP_071581.1 | 4 | 370 | ||
GenBank mRNA | AF007107.1 | - | 4 | 391 | ||
AF007108.1 | - | 4 | 333 | |||
BC086945.1 | - | 4 | 364 | 7322141 | ||
D13205.1 | - | 4 | 370 | |||
FQ209861.1 | - | 4 | 361 | |||
FQ210790.1 | - | 4 | 358 | |||
FQ211029.1 | - | 2 | 46 | |||
FQ224354.1 | - | 4 | 361 |
