siRNA Details
Gene Symbol: Clec10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022393.2 | NP_071788.2 | 8 | 878 | ||
XM_006246803.2 | XP_006246865.1 | 8 | 916 | |||
XM_008767867.1 | XP_008766089.1 | 8 | 916 | |||
XM_008767868.1 | XP_008766090.1 | 8 | 760 | |||
XM_008767869.1 | XP_008766091.1 | 8 | 757 | |||
GenBank mRNA | BC091151.1 | - | 8 | 878 | 7375234 | |
FQ213042.1 | - | 8 | 750 | |||
FQ228437.1 | - | 8 | 736 | |||
J05495.1 | - | 8 | 738 |
